Search
Search

Grb14| Gene Symbol: Grb14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016719.1 | NP_057928.1 | 7 - 8 | 1042 | 81 | |
| GenBank mRNA | AF155647.1 | - | 7 - 8 | 1042 | 81 | |
| AK147041.1 | - | 7 - 8 | 1051 | 81 | ||
| BC138581.1 | - | 7 - 8 | 948 | 8861601 | 81 | |
| BC145913.1 | - | 7 - 8 | 948 | 40131216 | 81 | |
Set Membership: |
|