Search
Search
| Gene Symbol: Wbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018765.3 | NP_061235.2 | 1 - 2 | 343 | 109 | |
| GenBank mRNA | AF071184.1 | - | 1 - 2 | 195 | 109 | |
| AK019160.1 | - | 1 - 2 | 77 | 109 | ||
| AK135215.1 | - | 1 - 2 | 343 | 109 | ||
| AK160559.1 | - | 1 - 2 | 80 | 109 | ||
| AK162009.1 | - | 1 - 2 | 85 | 109 | ||
Set Membership: |
|