Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00498260_m1
          See other KCND3 GE Assays ›
          Gene Symbol:
          Kcnd3
          Gene Name:
          potassium voltage-gated channel, Shal-related family, member 3
          Gene Aliases:
          AW045978, Kncd3, Kv4.3
          Chromosome Location:
          Chr.3: 105452093 - 105674002 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00498260_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kcnd3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019931.1 NP_064315.1 4 - 5 1459 84
          GenBank mRNA AF107781.1 - 4 - 5 1459 84
          BC141097.1 - 4 - 5 1493 9055709 84
          BC141098.1 - 4 - 5 1493 9055710 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300001092.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kcnd3

          Panther Classification:

          Molecular Function -

          voltage-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          ion transport
          potassium ion transport
          regulation of ion transmembrane transport
          protein homooligomerization
          transmembrane transport
          potassium ion export
          cellular response to BMP stimulus
          potassium ion transmembrane transport
          membrane repolarization
          ion channel activity
          voltage-gated ion channel activity
          voltage-gated potassium channel activity
          A-type (transient outward) potassium channel activity
          potassium channel activity
          protein binding
          ion channel binding
          metal ion binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0300008819.mm.1 NM_001039347.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300008819.mm.1 NM_001039347.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300008819.mm.1 TC0300001092.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300008821.mm.1 NM_001039347.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300008821.mm.1 NM_001039347.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300008821.mm.1 NM_019931.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300008821.mm.1 NM_019931.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300008821.mm.1 TC0300001092.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300008825.mm.1 NM_019931.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300008825.mm.1 NM_019931.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300008825.mm.1 TC0300001092.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300008820.mm.1 NM_001039347.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300008820.mm.1 NM_001039347.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300008820.mm.1 NM_019931.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300008820.mm.1 NM_019931.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300008820.mm.1 TC0300001092.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300008823.mm.1 NM_001039347.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300008823.mm.1 NM_001039347.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300008823.mm.1 NM_019931.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300008823.mm.1 NM_019931.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300008823.mm.1 TC0300001092.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300008822.mm.1 NM_001039347.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300008822.mm.1 NM_001039347.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300008822.mm.1 NM_019931.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300008822.mm.1 NM_019931.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300008822.mm.1 TC0300001092.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 757447 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline