Search
Search

Itgax| Gene Symbol: Itgax | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021334.2 | NP_067309.1 | 23 - 24 | 2872 | 94 | |
| GenBank mRNA | AF211864.1 | - | 23 - 24 | 2872 | 94 | |
| AK155816.1 | - | 23 - 24 | 2894 | 94 | ||
| AK170432.1 | - | 23 - 24 | 2890 | 94 | ||
Set Membership: |
|