Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00498802_m1
          See other 0610007P14RIK GE Assays ›
          Gene Symbol:
          0610007P14Rik
          Gene Name:
          RIKEN cDNA 0610007P14 gene
          Gene Aliases:
          1190004E09Rik, AU019315, C77855, ORF11
          Chromosome Location:
          Chr.12: 85815447 - 85824921 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00498802_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 0610007P14Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021446.2 NP_067421.1 1 - 2 73 93
          GenBank mRNA AF270646.1 - 1 - 2 72 92
          AK004480.1 - 1 - 2 95 93
          AK156525.1 - 1 - 2 73 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200002101.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AF270646.1,AK004480.1,NM_021446.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 0610007P14Rik

          Gene Ontology Categories:

          Function(s)

          lipid metabolic process
          steroid biosynthetic process
          steroid metabolic process
          sterol biosynthetic process

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28606266 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28606268 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28606271 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28606270 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P400375 AF270646.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P400375 AK004480.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P400375 AK152230.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P400375 BC004591.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P400375 NM_021446.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P400375 AF270646.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P400375 AK004480.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P400375 AK152230.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P400375 BC004591.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P400375 NM_021446.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 308190 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 263154 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 568522 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 879877 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 391331 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 58953 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P400375 AF270646.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P400375 AK004480.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P400375 AK152230.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P400375 BC004591.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P400375 NM_021446.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P400375 AF270646.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P400375 AK004480.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P400375 AK152230.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P400375 BC004591.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P400375 NM_021446.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28606286 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28606290 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28606302 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28606294 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28606288 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28606303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28606270 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28606251 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28606253 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28606278 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28606280 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28606283 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28606265 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28606260 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28606298 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28606273 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28606266 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28606276 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28606271 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28606264 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28606299 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28606295 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28606262 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28606256 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28606269 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28606272 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28606255 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28606301 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28606259 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28606291 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28606252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28606285 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28606258 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28606296 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28606254 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28606267 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02568476 - -
          Whole Mouse Genome G4122A A_51_P400375 AF270646.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400375 AK004480.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400375 AK152230.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400375 BC004591.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400375 NM_021446.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400375 AF270646.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400375 AK004480.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400375 AK152230.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400375 BC004591.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400375 NM_021446.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113230 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02568476 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09995118 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28606266 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28606271 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28606270 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28606268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28606218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28606266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28606223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28606212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28606265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28606211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28606229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28606258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28606262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28606252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28606216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28606236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28606204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28606235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28606234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28606222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28606251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28606230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28606201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28606202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28606255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28606199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28606241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28606233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28606228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28606254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28606214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28606259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28606237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28606225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28606245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28606249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28606205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28606260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28606232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28606239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28606248 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10098808 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28606225 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28606239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10098814 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28606219 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28606248 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28606254 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28606257 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28606260 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28606265 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28606251 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28606228 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10421294 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28606232 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10148130 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09746677 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28606252 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28606255 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28606235 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28606249 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28606259 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28606220 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28606234 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28606266 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28606223 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10083201 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10421291 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28606264 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28606230 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28606221 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28606236 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10401671 - -
          Affymetrix Mouse Gene 1.0 ST Array 10401668 - -
          Affymetrix Mouse Gene 1.0 ST Array 10401669 - -
          Affymetrix Mouse Gene 1.0 ST Array 10401670 - -
          Affymetrix Mouse Gene 1.0 ST Array 10401672 - -
          Affymetrix HT MG-430 PM Array Plate 1454161_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10401667 - -
          Mouse Genome 430 2.0 Array 1454161_PM_s_at - -
          Mouse Genome 430 2.0 Array 1458286_PM_at - -
          Mouse Genome 430 2.0 Array 1454161_s_at - -
          Mouse Genome 430 2.0 Array 1458286_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 58520_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 58520_at - -
          Affymetrix Mouse Clariom S Array PSR1200014570.mm.1 NM_021446.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014570.mm.1 NM_021446.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014570.mm.1 TC1200002101.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200014573.mm.1 NM_021446.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014573.mm.1 NM_021446.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014573.mm.1 TC1200002101.mm.1 MTA TC
          Affymetrix HT Mouse Genome 430A Array 1454161_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1454161_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 795835 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1240685 - -
          MouseWG-6 v2 Expression BeadChip 6450273 - -
          Illumina mouse-6 v1.0 expression beadchip 1240685 - -
          Illumina mouse-6 v1.0 expression beadchip 6450273 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001240685 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006450273 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2773981 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2771349 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2773981 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2771349 - -
          mouse-6 v1.1 (Illumina) scl058520.1_161-S - -
          mouse-6 v1.1 (Illumina) scl42219.1_29-S - -
          Illumina Sentrix mouse-6.v1 bead array 6450273 - -
          Illumina Sentrix mouse-6.v1 bead array 1240685 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl058520.1_161-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl42219.1_29-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1240685 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6450273 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2808939 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2771349 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2808939 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2773981 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl058520.1_161-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl42219.1_29-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline