Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00499156_m1
          See other GPM6B GE Assays ›
          Gene Symbol:
          Gpm6b
          Gene Name:
          glycoprotein m6b
          Gene Aliases:
          AI593561, Gpm6, M6B
          Chromosome Location:
          Chr.X: 166238943 - 166389033 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00499156_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gpm6b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001177955.1 NP_001171426.1 3 - 4 834 108
          NM_001177956.1 NP_001171427.1 3 - 4 834 108
          NM_001177957.1 NP_001171428.1 3 - 4 834 108
          NM_001177958.1 NP_001171429.1 2 - 3 714 108
          NM_001177959.1 NP_001171430.1 2 - 3 714 108
          NM_001177960.1 NP_001171431.1 2 - 3 714 108
          NM_001177961.1 NP_001171432.1 2 - 3 379 108
          NM_001177962.1 NP_001171433.1 2 - 3 379 108
          NM_023122.3 NP_075611.1 2 - 3 379 108
          GenBank mRNA AF254869.1 - 2 - 3 575 108
          AF254870.1 - 3 - 4 695 108
          AF254871.1 - 2 - 3 350 108
          AF254872.1 - 2 - 3 575 108
          AF254873.1 - 2 - 3 575 108
          AF254874.1 - 3 - 4 695 108
          AF254875.1 - 3 - 4 695 108
          AF254876.1 - 2 - 3 350 108
          AF254877.1 - 2 - 3 350 108
          AF254878.1 - 3 - 4 722 108
          AF254879.1 - 4 - 5 842 108
          AK016567.1 - 2 - 3 304 108
          AK132778.1 - 2 - 3 366 108
          AK140714.1 - 2 - 3 394 108
          BC003912.1 - 1 - 2 162 108
          BC025165.1 - 2 - 3 340 3981536 108
          S65737.1 - 2 - 3 371 108

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00001682.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gpm6b

          Panther Classification:

          Molecular Function -

          myelin protein

          Gene Ontology Categories:

          Function(s)

          ossification
          transport
          protein transport
          positive regulation of bone mineralization
          regulation of actin cytoskeleton organization
          negative regulation of serotonin uptake
          regulation of focal adhesion assembly
          extracellular matrix assembly
          negative regulation of protein localization to cell surface

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0X00010790.mm.1 NM_001177955.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00010790.mm.1 NM_001177955.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00010790.mm.1 NM_001177956.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00010790.mm.1 NM_001177956.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00010790.mm.1 NM_001177957.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00010790.mm.1 NM_001177957.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00010790.mm.1 NM_001177958.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00010790.mm.1 NM_001177958.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00010790.mm.1 NM_001177959.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00010790.mm.1 NM_001177959.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00010790.mm.1 NM_001177960.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00010790.mm.1 NM_001177960.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00010790.mm.1 NM_001177961.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00010790.mm.1 NM_001177961.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00010790.mm.1 NM_001177962.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00010790.mm.1 NM_001177962.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00010790.mm.1 NM_023122.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00010790.mm.1 NM_023122.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00010790.mm.1 TC0X00001682.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline