| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0300013394.mm.1 |
NM_001146324.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300013394.mm.1 |
NM_001146324.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300013394.mm.1 |
NM_080634.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300013394.mm.1 |
NM_080634.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300013394.mm.1 |
TC0300001701.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300013393.mm.1 |
NM_001146324.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300013393.mm.1 |
NM_001146324.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300013393.mm.1 |
NM_080634.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300013393.mm.1 |
NM_080634.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300013393.mm.1 |
TC0300001701.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300013392.mm.1 |
NM_001146323.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300013392.mm.1 |
NM_001146323.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300013392.mm.1 |
NM_001146324.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300013392.mm.1 |
NM_001146324.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300013392.mm.1 |
NM_080634.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300013392.mm.1 |
NM_080634.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300013392.mm.1 |
TC0300001701.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300013396.mm.1 |
NM_001146323.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300013396.mm.1 |
NM_001146323.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300013396.mm.1 |
NM_001146324.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300013396.mm.1 |
NM_001146324.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300013396.mm.1 |
NM_080634.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300013396.mm.1 |
NM_080634.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300013396.mm.1 |
TC0300001701.mm.1 |
MTA TC |