| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300000072.mm.1 |
NM_001267724.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300000072.mm.1 |
NM_178283.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300000072.mm.1 |
TC1300000009.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300000077.mm.1 |
NM_001267724.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300000077.mm.1 |
NM_178283.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300000077.mm.1 |
TC1300000009.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300000079.mm.1 |
NM_178283.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300000079.mm.1 |
TC1300000009.mm.1 |
MTA TC |