Search
Search

Cdhr1| Gene Symbol: Cdhr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130878.2 | NP_570948.1 | 13 - 14 | 1658 | 64 | |
| GenBank mRNA | AB093300.1 | - | 13 - 14 | 2074 | 64 | |
| AF426393.1 | - | 13 - 14 | 1545 | 64 | ||
| AK143151.1 | - | 13 - 14 | 1599 | 64 | ||
| BC042454.1 | - | 13 - 14 | 1568 | 5369576 | 64 | |
| BC058270.1 | - | 13 - 14 | 1658 | 6490597 | 64 | |
Set Membership: |
|