Search
Search
| Gene Symbol: Pcx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162946.1 | NP_001156418.1 | 3 - 4 | 287 | 81 | |
| NM_008797.3 | NP_032823.2 | 2 - 3 | 264 | 81 | ||
| GenBank mRNA | AK128927.1 | - | 214 | 81 | ||
| AK148328.1 | - | 3 - 4 | 234 | 81 | ||
| AK170596.1 | - | 3 - 4 | 247 | 81 | ||
| AK170871.1 | - | 3 - 4 | 237 | 81 | ||
| AK172320.1 | - | 3 - 4 | 215 | 81 | ||
| BC055030.1 | - | 3 - 4 | 222 | 5053475 | 81 | |
| BG087228.2 | - | 3 - 4 | 223 | 81 | ||
| BY139237.1 | - | 2 - 3 | 81 | |||
| BY230152.1 | - | 3 - 4 | 81 | |||
| L09192.1 | - | 2 - 3 | 240 | 81 | ||
Set Membership: |
|