Search
Search

Plxna3| Gene Symbol: Plxna3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008883.2 | NP_032909.2 | 12 - 13 | 2525 | 98 | |
| GenBank mRNA | BC093482.1 | - | 12 - 13 | 2508 | 30945372 | 98 |
| D86950.1 | - | 12 - 13 | 2491 | 98 | ||
Set Membership: |
|