Search
Search

Creb1| Gene Symbol: Creb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009952.2 | NP_034082.1 | 2 - 3 | 335 | 68 | |
| NM_133828.2 | NP_598589.2 | 2 - 3 | 335 | 68 | ||
| GenBank mRNA | AF448507.1 | - | 2 - 3 | 211 | 68 | |
| AF448508.1 | - | 863 | 68 | |||
| AK042595.1 | - | 2 - 3 | 308 | 68 | ||
| AK044992.1 | - | 2 - 3 | 150 | 68 | ||
| AK044993.1 | - | 2 - 3 | 150 | 68 | ||
| AK079223.1 | - | 2 - 3 | 331 | 68 | ||
| BC021649.1 | - | 2 - 3 | 268 | 4017170 | 68 | |
| BY782293.1 | - | 2 - 3 | 292 | 68 | ||
| M95106.1 | - | 2 - 3 | 245 | 68 | ||
| U46027.1 | - | 1 - 2 | 83 | 68 | ||
Set Membership: |
|