Search
Search
| Gene Symbol: Cib1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291275.1 | NP_001278204.1 | 1 - 2 | 292 | 83 | |
| NM_011870.5 | NP_036000.1 | 1 - 2 | 292 | 83 | ||
| GenBank mRNA | AB006463.1 | - | 1 - 2 | 73 | 83 | |
| AF173010.1 | - | 1 - 2 | 71 | 83 | ||
| AK010345.1 | - | 1 - 2 | 89 | 83 | ||
| AK027915.1 | - | 1 - 2 | 104 | 83 | ||
| AK152805.1 | - | 1 - 2 | 124 | 83 | ||
| AK165472.1 | - | 1 - 2 | 77 | 83 | ||
| BC003714.1 | - | 1 - 2 | 78 | 3599986 | 83 | |
| BC054385.1 | - | 1 - 2 | 97 | 6476137 | 83 | |
| CF104297.1 | - | 1 - 2 | 300 | 83 | ||
Set Membership: |
|