Search
Search

Gtf2h2| Gene Symbol: Gtf2h2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022011.4 | NP_071294.3 | 2 - 3 | 242 | 110 | |
| GenBank mRNA | AB221674.1 | - | 1 - 2 | 56 | 110 | |
| AK010543.1 | - | 2 - 3 | 213 | 110 | ||
| AK078297.1 | - | 2 - 3 | 213 | 110 | ||
| AK145720.1 | - | 2 - 3 | 222 | 110 | ||
| AK154739.1 | - | 2 - 3 | 216 | 110 | ||
| BC016231.1 | - | 2 - 3 | 197 | 4488798 | 110 | |
| BC053382.1 | - | 2 - 3 | 206 | 30047823 | 110 | |
| BY277671.1 | - | 2 - 3 | 242 | 110 | ||
Set Membership: |
|