Search
Search
| Gene Symbol: Pecr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023523.5 | NP_076012.3 | 4 - 5 | 606 | 108 | |
| GenBank mRNA | AF232011.1 | - | 4 - 5 | 512 | 108 | |
| AF242204.1 | - | 4 - 5 | 546 | 108 | ||
| AK010260.1 | - | 4 - 5 | 597 | 108 | ||
| AK090327.1 | - | 4 - 5 | 535 | 108 | ||
| AK135714.1 | - | 4 - 5 | 551 | 108 | ||
| AK162100.1 | - | 4 - 5 | 554 | 109 | ||
| BC013530.1 | - | 4 - 5 | 530 | 4238679 | 108 | |
Set Membership: |
|