Search
Search

Hvcn1| Gene Symbol: Hvcn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042489.2 | NP_001035954.1 | 4 - 5 | 651 | 65 | |
| NM_028752.3 | NP_083028.1 | 3 - 4 | 513 | 65 | ||
| GenBank mRNA | AK002854.1 | - | 3 - 4 | 514 | 65 | |
| AK154880.1 | - | 4 - 5 | 543 | 65 | ||
| AK155123.1 | - | 4 - 5 | 488 | 65 | ||
| AK170422.1 | - | 4 - 5 | 512 | 65 | ||
| BC021548.1 | - | 4 - 5 | 482 | 4913027 | 65 | |
| BY703352.1 | - | 3 - 4 | 507 | 65 | ||
| CF734112.1 | - | 4 - 5 | 486 | 65 | ||
| CF749152.1 | - | 4 - 5 | 651 | 65 | ||
Set Membership: |
|