Search
Search

Slamf9| Gene Symbol: Slamf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029612.4 | NP_083888.3 | 1 - 2 | 185 | 66 | |
| GenBank mRNA | AB196824.1 | - | 1 - 2 | 48 | 66 | |
| AB196825.1 | - | 1 - 2 | 48 | 66 | ||
| AB196826.1 | - | 1 - 2 | 48 | 66 | ||
| AB196827.1 | - | 1 - 2 | 48 | 66 | ||
| AI120415.1 | - | 1 - 2 | 188 | 66 | ||
| AK009505.1 | - | 1 - 2 | 141 | 66 | ||
| AK090041.1 | - | 1 - 2 | 176 | 66 | ||
| BC019477.1 | - | 1 - 2 | 124 | 4458767 | 66 | |
Set Membership: |
|