Search
Search

Gpnmb| Gene Symbol: Gpnmb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053110.4 | NP_444340.3 | 4 - 5 | 750 | 64 | |
| GenBank mRNA | AF322054.1 | - | 4 - 5 | 588 | 64 | |
| AJ251685.1 | - | 4 - 5 | 635 | 64 | ||
| AK044764.1 | - | 4 - 5 | 644 | 64 | ||
| AK076347.1 | - | 4 - 5 | 619 | 64 | ||
| AK079220.1 | - | 592 | 64 | |||
| AK149700.1 | - | 4 - 5 | 619 | 64 | ||
| AK149740.1 | - | 4 - 5 | 657 | 64 | ||
| AK150739.1 | - | 4 - 5 | 619 | 64 | ||
| AK154617.1 | - | 4 - 5 | 654 | 64 | ||
| AK159165.1 | - | 4 - 5 | 649 | 64 | ||
| AK159270.1 | - | 4 - 5 | 657 | 64 | ||
| AK159747.1 | - | 4 - 5 | 619 | 64 | ||
| AK159908.1 | - | 4 - 5 | 625 | 64 | ||
| AK170506.1 | - | 4 - 5 | 649 | 64 | ||
| AK171441.1 | - | 4 - 5 | 619 | 64 | ||
| AK171465.1 | - | 4 - 5 | 619 | 64 | ||
| AK171474.1 | - | 4 - 5 | 619 | 64 | ||
| AK171517.1 | - | 4 - 5 | 737 | 64 | ||
| AK171608.1 | - | 4 - 5 | 619 | 64 | ||
| AK171618.1 | - | 4 - 5 | 619 | 64 | ||
| BC026375.1 | - | 4 - 5 | 624 | 4164706 | 64 | |
Set Membership: |
|