Search
Search
| Gene Symbol: Ric8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053194.4 | NP_444424.1 | 2 - 3 | 207 | 75 | |
| GenBank mRNA | AF288813.1 | - | 2 - 3 | 206 | 75 | |
| AK080192.1 | - | 2 - 3 | 150 | 75 | ||
| AK155282.1 | - | 2 - 3 | 182 | 75 | ||
| AK165536.1 | - | 2 - 3 | 184 | 75 | ||
| AK167746.1 | - | 2 - 3 | 210 | 75 | ||
| AK169367.1 | - | 2 - 3 | 171 | 75 | ||
| AK212668.1 | - | 1 - 2 | 55 | 75 | ||
| BC022917.1 | - | 2 - 3 | 184 | 4460521 | 75 | |
Set Membership: |
|