Search
Search

Srgap3| Gene Symbol: Srgap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080448.4 | NP_536696.4 | 13 - 14 | 2374 | 102 | |
| GenBank mRNA | AF338472.1 | - | 12 - 13 | 1532 | 102 | |
| AF496547.1 | - | 13 - 14 | 1604 | 102 | ||
| AK122276.1 | - | 13 - 14 | 1950 | 102 | ||
| CK032058.1 | - | 4 - 5 | 265 | 102 | ||
Set Membership: |
|