Search
Search

Idh3b| Gene Symbol: Idh3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130884.4 | NP_570954.1 | 3 - 4 | 247 | 75 | |
| GenBank mRNA | AK046059.1 | - | 2 - 3 | 367 | 75 | |
| AY035212.1 | - | 3 - 4 | 228 | 75 | ||
| BC009022.1 | - | 3 - 4 | 236 | 3257667 | 75 | |
| BY336999.1 | - | 3 - 4 | 247 | 75 | ||
| DN176034.1 | - | 3 - 4 | 213 | 75 | ||
Set Membership: |
|