Search
Search

Ppargc1b| Gene Symbol: Ppargc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133249.2 | NP_573512.1 | 8 - 9 | 2656 | 138 | |
| GenBank mRNA | AF453324.1 | - | 8 - 9 | 2656 | 138 | |
| AK028464.1 | - | 9 - 10 | 2706 | 138 | ||
| AK086111.1 | - | 8 - 9 | 2672 | 138 | ||
| BC150699.1 | - | 8 - 9 | 2636 | 9087610 | 138 | |
Set Membership: |
|