Search
Search

Atp6v1h| Gene Symbol: Atp6v1h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133826.4 | NP_598587.2 | 3 - 4 | 354 | 90 | |
| GenBank mRNA | AK034165.1 | - | 3 - 4 | 405 | 90 | |
| AK078767.1 | - | 3 - 4 | 355 | 90 | ||
| AK081492.1 | - | 3 - 4 | 364 | 90 | ||
| AK151659.1 | - | 3 - 4 | 417 | 90 | ||
| AK152147.1 | - | 3 - 4 | 375 | 90 | ||
| BC009154.1 | - | 3 - 4 | 355 | 3601621 | 90 | |
Set Membership: |
|