| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200005126.mm.1 |
NM_001080968.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005126.mm.1 |
NM_001080968.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005126.mm.1 |
NM_133852.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005126.mm.1 |
NM_133852.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005126.mm.1 |
TC0200000574.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200005128.mm.1 |
NM_001080968.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005128.mm.1 |
NM_001080968.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005128.mm.1 |
NM_133852.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005128.mm.1 |
NM_133852.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005128.mm.1 |
TC0200000574.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200005124.mm.1 |
NM_001080968.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005124.mm.1 |
NM_001080968.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005124.mm.1 |
NM_133852.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005124.mm.1 |
NM_133852.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005124.mm.1 |
TC0200000574.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200005125.mm.1 |
NM_001080968.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005125.mm.1 |
NM_001080968.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005125.mm.1 |
NM_133852.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005125.mm.1 |
NM_133852.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005125.mm.1 |
TC0200000574.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200005129.mm.1 |
NM_001080968.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005129.mm.1 |
NM_001080968.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005129.mm.1 |
NM_133852.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005129.mm.1 |
NM_133852.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005129.mm.1 |
TC0200000574.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200005130.mm.1 |
NM_001080968.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005130.mm.1 |
NM_001080968.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005130.mm.1 |
NM_133852.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005130.mm.1 |
NM_133852.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005130.mm.1 |
TC0200000574.mm.1 |
MTA TC |