Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00506343_m1
          See other AKR1C14 GE Assays ›
          Gene Symbol:
          Akr1c14
          Gene Name:
          aldo-keto reductase family 1, member C14
          Gene Aliases:
          9030611N15Rik, AW557061
          Chromosome Location:
          Chr.13: 4048999 - 4090560 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00506343_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Akr1c14
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134072.1 NP_598833.1 8 - 9 974 100
          GenBank mRNA AK033537.1 - 8 - 9 992 100
          AK136528.1 - 8 - 9 991 100
          AY730283.1 - 8 - 9 936 100
          BC013482.1 - 8 - 9 974 4240772 100
          BC026628.1 - 8 - 9 973 4983237 100

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300000016.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Akr1c14

          Panther Classification:

          Molecular Function -

          oxidoreductase reductase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          oxidation-reduction process
          steroid dehydrogenase activity
          androsterone dehydrogenase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2007155 AK136528.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2007155 AK033537.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 236908 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 503452 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 354396 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 571488 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 465461 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 970342 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 139122 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 64359 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 378146 BC104078.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 378146 BC104079.2 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 378146 NM_001011525.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 378146 NM_147038.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 378146 NM_147039.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 643269 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 227746 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 836034 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 170795 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 163239 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 44312 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P132955 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2007155 AK136528.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2007155 AK033537.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791134 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791117 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791139 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791132 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791137 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791108 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791136 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791144 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791133 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791115 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791109 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791123 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791126 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791129 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791121 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791124 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791107 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791128 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791119 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791130 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791141 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791105 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791113 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07771723 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P132955 - -
          Whole Mouse Genome G4122A A_51_P284177 BC026628.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P284177 AK136528.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P284177 BC013482.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P284177 NM_134072.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P284177 AK033537.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126285 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07771725 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P132955 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10580051 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10580045 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10580043 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10580056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791141 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791138 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791118 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791091 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10580045 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10469331 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791157 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10966592 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791168 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791105 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791166 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791130 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791150 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791089 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791128 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791148 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791154 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791147 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791092 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791094 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791119 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791153 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791090 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791095 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791158 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791080 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791097 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791101 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791077 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791109 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10580056 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791081 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791141 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791085 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791115 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791121 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11087980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791124 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791151 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791139 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791113 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791123 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791103 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791099 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791144 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10543002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791083 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791146 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791082 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791133 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791108 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791162 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11047174 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791134 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791160 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10403293 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403302 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403300 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403301 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403297 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403294 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403296 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403299 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403295 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403292 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403298 - -
          Affymetrix HT MG-430 PM Array Plate 1418979_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10403291 - -
          Mouse Genome 430 2.0 Array 1418979_at - -
          Mouse Genome 430 2.0 Array 1418979_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 105387_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 105387_at - -
          Affymetrix Mouse Clariom S Array PSR1300000125.mm.1 NM_134072.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000125.mm.1 NM_134072.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000125.mm.1 TC1300000016.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300000124.mm.1 NM_134072.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000124.mm.1 NM_134072.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000124.mm.1 TC1300000016.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300000126.mm.1 NM_134072.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000126.mm.1 NM_134072.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000126.mm.1 TC1300000016.mm.1 MTA TC
          Mouse Expression Set 430 1418979_PM_at - -
          Mouse Expression Set 430 1418979_at - -
          Mouse Genome 430A 2.0 Array 1418979_at - -
          Mouse Genome 430A 2.0 Array 1418979_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418979_at - -
          Affymetrix HT Mouse Genome 430A Array 1418979_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 524587 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2370270 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002370270 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2655871 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2655871 - -
          mouse-6 v1.1 (Illumina) scl45103.13_79-S - -
          Illumina Sentrix mouse-6.v1 bead array 2370270 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl45103.13_79-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2370270 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2655871 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2921215 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2655871 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2921215 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline