Search
Search

Slc12a8| Gene Symbol: Slc12a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083902.1 | NP_001077371.1 | 6 - 7 | 846 | 75 | |
| NM_134251.2 | NP_599012.2 | 6 - 7 | 815 | 75 | ||
| GenBank mRNA | AF319951.1 | - | 6 - 7 | 809 | 75 | |
| AK087781.1 | - | 2 - 3 | 404 | 75 | ||
| AK165080.1 | - | 2 - 3 | 469 | 75 | ||
| BC030926.1 | - | 6 - 7 | 846 | 4910321 | 75 | |
Set Membership: |
|