Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00507514_m1
          See other APPL1 GE Assays ›
          Gene Symbol:
          Appl1
          Gene Name:
          adaptor protein, phosphotyrosine interaction, PH domain and leucine zipper containing 1
          Gene Aliases:
          2900057D21Rik, 7330406P05Rik, AI585782, AW209077, BB022931, C88264, DIP13
          Chromosome Location:
          Chr.14: 26918986 - 26971205 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00507514_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Appl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145221.2 NP_660256.1 3 - 4 317 92
          GenBank mRNA AK013715.1 - 3 - 4 314 92
          AK030445.1 - 3 - 4 197 92
          AK033566.1 - 3 - 4 288 92
          AK045438.1 - 3 - 4 253 92
          AK049307.1 - 3 - 4 304 92
          AK146356.1 - 3 - 4 288 92
          AY113705.1 - 3 - 4 282 92
          BC063751.1 - 3 - 4 329 6511401 92
          BU514763.1 - 3 - 4 328 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400001627.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Appl1

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein GTPase-activating protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cell cycle
          cell proliferation
          regulation of glucose import
          regulation of establishment of protein localization to plasma membrane
          identical protein binding
          protein kinase B binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1400012287.mm.1 NM_145221.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400012287.mm.1 NM_145221.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400012287.mm.1 TC1400001627.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400012289.mm.1 NM_145221.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400012289.mm.1 NM_145221.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400012289.mm.1 TC1400001627.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400012291.mm.1 NM_145221.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400012291.mm.1 NM_145221.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400012291.mm.1 TC1400001627.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400012286.mm.1 NM_145221.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400012286.mm.1 NM_145221.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400012286.mm.1 TC1400001627.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400012290.mm.1 NM_145221.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400012290.mm.1 NM_145221.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400012290.mm.1 TC1400001627.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400012288.mm.1 NM_145221.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400012288.mm.1 NM_145221.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400012288.mm.1 TC1400001627.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 456407 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline