Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00508295_m1
          See other 1810037I17RIK GE Assays ›
          Gene Symbol:
          1810037I17Rik
          Gene Name:
          RIKEN cDNA 1810037I17 gene
          Gene Aliases:
          5730414I01Rik
          Chromosome Location:
          Chr.3: 122924397 - 122926194 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00508295_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1810037I17Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024461.2 NP_077781.1 1 - 2 264 68
          GenBank mRNA AK007721.1 - 1 - 2 265 68
          AK009748.1 - 1 - 2 230 68
          AK013091.1 - 1 - 2 249 68
          AK170776.1 - 1 - 2 230 68
          BC002135.1 - 1 - 2 273 3484100 68
          BC086913.1 - 1 - 2 229 30121587 68
          BC091739.1 - 1 - 2 232 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300001252.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 1810037I17Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2044593 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P303176 AK170776.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P303176 BC091739.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P303176 NM_024461.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P303176 AK170776.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P303176 BC091739.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P303176 NM_024461.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2132207 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 150036 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2044593 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P303176 AK170776.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P303176 BC091739.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P303176 NM_024461.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P303176 AK170776.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P303176 BC091739.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P303176 NM_024461.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2132207 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09561939 - -
          Whole Mouse Genome G4122A A_51_P217967 AK170776.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P217967 BC002135.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P217967 BC091739.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P217967 NM_024461.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P217967 AK007721.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485187 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32976491 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02853009 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32976489 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32976470 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32976492 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32976472 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485184 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485193 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02755060 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02759400 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485160 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485191 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32976483 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485188 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485178 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485164 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32976486 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02853008 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485163 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32976473 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32976475 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485183 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32976478 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485165 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485179 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485168 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32976477 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485181 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02749740 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02749262 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485173 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32976474 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485167 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485187 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10495828 - -
          Affymetrix Mouse Gene 1.0 ST Array 10604020 - -
          Affymetrix Mouse Gene 1.0 ST Array 10495827 - -
          Affymetrix Mouse Gene 1.0 ST Array 10495829 - -
          Affymetrix HT MG-430 PM Array Plate 1424365_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10495826 - -
          Affymetrix Mouse Gene 1.1 ST Array 10604019 - -
          Mouse Genome 430 2.0 Array 1424365_at - -
          Mouse Genome 430 2.0 Array 1424365_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67704_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67704_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0300010234.mm.1 NM_024461.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300010234.mm.1 NM_024461.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300010234.mm.1 TC0300001252.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300010238.mm.1 NM_024461.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300010238.mm.1 NM_024461.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300010238.mm.1 TC0300001252.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300010236.mm.1 NM_024461.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300010236.mm.1 NM_024461.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300010236.mm.1 TC0300001252.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300010235.mm.1 NM_024461.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300010235.mm.1 NM_024461.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300010235.mm.1 TC0300001252.mm.1 MTA TC
          Mouse Expression Set 430 1424365_PM_at - -
          Mouse Expression Set 430 1424365_at - -
          Murine Genome U74v2 Set 95406_PM_at - -
          Murine Genome U74v2 Set 102234_at - -
          Murine Genome U74v2 Set 95406_at - -
          Murine Genome U74v2 Set 102234_PM_at - -
          Mouse Genome 430A 2.0 Array 1424365_PM_at - -
          Mouse Genome 430A 2.0 Array 1424365_at - -
          Affymetrix HT Mouse Genome 430A Array 1424365_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424365_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 908428 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3610563 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003610563 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2602607 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2602607 - -
          mouse-6 v1.1 (Illumina) scl067704.2_8-S - -
          Illumina Sentrix mouse-6.v1 bead array 3610563 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl067704.2_8-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3610563 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2602607 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl067704.2_8-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline