Search
Search

Ogfod2| Gene Symbol: Ogfod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025671.2 | NP_079947.1 | 1 - 2 | 129 | 57 | |
| GenBank mRNA | AK004916.1 | - | 1 - 2 | 128 | 57 | |
| AK004943.1 | - | 1 - 2 | 130 | 57 | ||
| AK017497.1 | - | 1 - 2 | 57 | |||
| AK018534.1 | - | 1 - 2 | 130 | 57 | ||
| AK168946.1 | - | 1 - 2 | 138 | 57 | ||
| BC069872.1 | - | 1 - 2 | 132 | 4219293 | 57 | |
Set Membership: |
|