| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1800006258.mm.1 |
NM_001164421.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006258.mm.1 |
NM_001164421.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006258.mm.1 |
NM_001164423.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006258.mm.1 |
NM_001164423.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006258.mm.1 |
NM_025861.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006258.mm.1 |
NM_025861.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006258.mm.1 |
TC1800000852.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800006261.mm.1 |
NM_001164420.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006261.mm.1 |
NM_001164420.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006261.mm.1 |
NM_001164421.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006261.mm.1 |
NM_001164421.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006261.mm.1 |
NM_001164422.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006261.mm.1 |
NM_001164422.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006261.mm.1 |
NM_001164423.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006261.mm.1 |
NM_001164423.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006261.mm.1 |
NM_001164424.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006261.mm.1 |
NM_001164424.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006261.mm.1 |
NM_025861.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006261.mm.1 |
NM_025861.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006261.mm.1 |
TC1800000852.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800006247.mm.1 |
TC1800000852.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800006248.mm.1 |
TC1800000852.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800006249.mm.1 |
NM_001164420.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006249.mm.1 |
NM_001164420.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006249.mm.1 |
TC1800000852.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800006250.mm.1 |
NM_001164420.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006250.mm.1 |
NM_001164420.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006250.mm.1 |
TC1800000852.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800006263.mm.1 |
NM_001164420.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006263.mm.1 |
NM_001164420.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006263.mm.1 |
NM_001164421.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006263.mm.1 |
NM_001164421.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006263.mm.1 |
NM_001164422.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006263.mm.1 |
NM_001164422.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006263.mm.1 |
NM_001164423.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006263.mm.1 |
NM_001164423.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006263.mm.1 |
NM_001164424.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006263.mm.1 |
NM_001164424.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006263.mm.1 |
NM_025861.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006263.mm.1 |
NM_025861.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006263.mm.1 |
TC1800000852.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800006252.mm.1 |
NM_001164422.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006252.mm.1 |
NM_001164422.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006252.mm.1 |
NM_001164424.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006252.mm.1 |
NM_001164424.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006252.mm.1 |
TC1800000852.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800006246.mm.1 |
TC1800000852.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800006259.mm.1 |
NM_001164421.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006259.mm.1 |
NM_001164421.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006259.mm.1 |
NM_001164423.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006259.mm.1 |
NM_001164423.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006259.mm.1 |
NM_025861.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006259.mm.1 |
NM_025861.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006259.mm.1 |
TC1800000852.mm.1 |
MTA TC |