Search
Search

Ift27| Gene Symbol: Ift27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025931.2 | NP_080207.1 | 5 - 6 | 658 | 64 | |
| GenBank mRNA | AK011196.1 | - | 5 - 6 | 656 | 64 | |
| AK044278.1 | - | 2 - 3 | 1096 | 64 | ||
| BC017514.1 | - | 5 - 6 | 658 | 4502942 | 64 | |
Set Membership: |
|