Search
Search

Gramd3| Gene Symbol: Gramd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026240.2 | NP_080516.2 | 8 - 9 | 880 | 59 | |
| GenBank mRNA | AK018551.1 | - | 9 - 10 | 1130 | 59 | |
| AK018691.1 | - | 8 - 9 | 866 | 59 | ||
| AK038289.1 | - | 7 - 8 | 921 | 59 | ||
| AK161329.1 | - | 8 - 9 | 813 | 59 | ||
| AK164050.1 | - | 8 - 9 | 865 | 59 | ||
| BC057978.1 | - | 8 - 9 | 855 | 5044170 | 59 | |
Set Membership: |
|