Search
Search

Tbc1d23| Gene Symbol: Tbc1d23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026254.2 | NP_080530.2 | 13 - 14 | 1515 | 93 | |
| GenBank mRNA | AK015437.1 | - | 4 - 5 | 347 | 93 | |
| AK155142.1 | - | 13 - 14 | 1536 | 93 | ||
| BC031706.1 | - | 13 - 14 | 1515 | 4194266 | 93 | |
Set Membership: |
|