Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00509512_s1
          See other ASPRV1 GE Assays ›
          Gene Symbol:
          Asprv1
          Gene Name:
          aspartic peptidase, retroviral-like 1
          Gene Aliases:
          2300003P22Rik, AA986851, SASP, SASPase, Taps
          Chromosome Location:
          Chr.6: 86628174 - 86629710 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00509512_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Asprv1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026414.2 NP_080690.2 1 601 73
          GenBank mRNA AK004007.1 - 1 48 73
          AK160494.1 - 1 543 73
          BC057938.1 - 1 614 73
          BC108357.1 - 1 499 73
          BC119099.1 - 1 117 73
          BC119101.1 - 1 117 73
          DQ841260.1 - 1 601 73

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Asprv1

          Panther Classification:

          Molecular Function -

          viral or transposable element protein

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          protein processing
          skin development
          aspartic-type endopeptidase activity
          peptidase activity
          hydrolase activity
          aspartic endopeptidase activity, intramembrane cleaving

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P499698 BC057938.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P499698 BC108357.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P499698 BC119099.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P499698 BC119101.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P499698 DQ841260.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P499698 BC057938.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P499698 BC108357.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P499698 BC119099.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P499698 BC119101.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P499698 DQ841260.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140381 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P499698 BC057938.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P499698 BC108357.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P499698 BC119099.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P499698 BC119101.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P499698 DQ841260.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P499698 BC057938.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P499698 BC108357.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P499698 BC119099.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P499698 BC119101.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P499698 DQ841260.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140381 - -
          Whole Mouse Genome G4122A A_51_P499698 BC057938.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P499698 BC108357.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P499698 BC119099.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P499698 BC119101.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P499698 DQ841260.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P499698 BC057938.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P499698 BC108357.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P499698 BC119099.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P499698 BC119101.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P499698 DQ841260.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137920 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140381 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05740590 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05740610 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05740594 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05051246 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05740602 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05740589 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05740616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24611087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05594382 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611105 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05631521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05022663 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05542330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611104 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05631523 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05740612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05740597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05051246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611117 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611051 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611063 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05591709 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611058 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05749355 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05749357 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611059 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611110 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05299028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05542328 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05051247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611111 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611098 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611099 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611121 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05749356 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611077 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05631530 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611113 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611038 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611102 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611043 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05740617 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611120 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611089 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24611056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05412939 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05740610 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10539740 - -
          Affymetrix HT MG-430 PM Array Plate 1452732_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10539739 - -
          Mouse Genome 430 2.0 Array 1452732_PM_at - -
          Mouse Genome 430 2.0 Array 1452732_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67855_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67855_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600007765.mm.1 NM_026414.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600007765.mm.1 NM_026414.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600007765.mm.1 TC0600000995.mm.1 MTA TC
          Mouse Expression Set 430 1452732_PM_at - -
          Mouse Expression Set 430 1452732_at - -
          Murine Genome U74v2 Set 95740_at - -
          Murine Genome U74v2 Set 95740_PM_at - -
          Mouse Genome 430A 2.0 Array 1452732_at - -
          Mouse Genome 430A 2.0 Array 1452732_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1452732_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1452732_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 704799 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2650079 - -
          Illumina mouse-6 v1.0 expression beadchip 2650079 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002650079 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2699923 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2699923 - -
          mouse-6 v1.1 (Illumina) scl29796.1.1_131-S - -
          Illumina Sentrix mouse-6.v1 bead array 2650079 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl29796.1.1_131-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2650079 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2699923 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2699923 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl29796.1.1_131-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline