Search
Search
| Gene Symbol: Dph5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027193.2 | NP_081469.2 | 3 - 4 | 379 | 87 | |
| GenBank mRNA | AK010475.1 | - | 3 - 4 | 369 | 87 | |
| AK034652.1 | - | 389 | 87 | |||
| AK045127.1 | - | 3 - 4 | 399 | 87 | ||
| AK045548.1 | - | 3 - 4 | 369 | 87 | ||
| AK046024.1 | - | 3 - 4 | 369 | 87 | ||
| AK046753.1 | - | 3 - 4 | 397 | 87 | ||
| BC060372.1 | - | 3 - 4 | 379 | 6391546 | 87 | |
Set Membership: |
|