Search
Search

Prpf3| Gene Symbol: Prpf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027541.4 | NP_081817.2 | 4 - 5 | 907 | 60 | |
| GenBank mRNA | AK014398.1 | - | 4 - 5 | 515 | 60 | |
| AK088194.1 | - | 4 - 5 | 908 | 60 | ||
| AK167435.1 | - | 4 - 5 | 544 | 60 | ||
| BC006782.1 | - | 4 - 5 | 492 | 3589226 | 60 | |
| BC026607.1 | - | 1 - 2 | 661 | 60 | ||
| BC040787.1 | - | 1 - 2 | 661 | 60 | ||
Set Membership: |
|