Search
Search

Spata16| Gene Symbol: Spata16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027583.3 | NP_081859.1 | 3 - 4 | 887 | 61 | |
| NM_029150.4 | NP_083426.1 | 3 - 4 | 887 | 61 | ||
| GenBank mRNA | AK014869.1 | - | 3 - 4 | 851 | 61 | |
| AK076628.1 | - | 3 - 4 | 888 | 61 | ||
| BC100443.1 | - | 3 - 4 | 868 | 30920480 | 61 | |
| BC132205.1 | - | 3 - 4 | 811 | 40130482 | 61 | |
| BC132539.1 | - | 3 - 4 | 811 | 40130816 | 61 | |
Set Membership: |
|