Search
Search

Spata1| Gene Symbol: Spata1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027617.3 | NP_081893.1 | 4 - 5 | 491 | 125 | |
| GenBank mRNA | BC139279.1 | - | 3 - 4 | 404 | 8862301 | 125 |
| BC139280.1 | - | 3 - 4 | 404 | 8862302 | 125 | |
| BC171970.1 | - | 3 - 4 | 404 | 125 | ||
Set Membership: |
|