Search
Search

Afmid| Gene Symbol: Afmid | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027827.3 | NP_082103.1 | 2 - 3 | 178 | 75 | |
| GenBank mRNA | AF399717.1 | - | 2 - 3 | 173 | 75 | |
| AY099479.1 | - | 2 - 3 | 173 | 75 | ||
| BC024452.1 | - | 2 - 3 | 149 | 75 | ||
| BC043309.1 | - | 2 - 3 | 144 | 75 | ||
| BC066780.1 | - | 2 - 3 | 157 | 30301832 | 75 | |
| BY780917.1 | - | 2 - 3 | 178 | 75 | ||
Set Membership: |
|