| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800007316.mm.1 |
NM_001168244.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800007316.mm.1 |
NM_001168244.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800007316.mm.1 |
NM_001168246.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800007316.mm.1 |
NM_001168246.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800007316.mm.1 |
NM_027954.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800007316.mm.1 |
NM_027954.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800007316.mm.1 |
NR_031759.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800007316.mm.1 |
NR_031759.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800007316.mm.1 |
TC0800000968.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800007315.mm.1 |
NM_001168244.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800007315.mm.1 |
NM_001168244.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800007315.mm.1 |
NM_001168246.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800007315.mm.1 |
NM_001168246.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800007315.mm.1 |
NM_027954.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800007315.mm.1 |
NM_027954.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800007315.mm.1 |
NR_031759.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800007315.mm.1 |
NR_031759.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800007315.mm.1 |
TC0800000968.mm.1 |
MTA TC |