Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00511710_m1
          See other TMEM138 GE Assays ›
          Gene Symbol:
          Tmem138
          Gene Name:
          transmembrane protein 138
          Gene Aliases:
          1700113I01Rik, 2900055D14Rik
          Chromosome Location:
          Chr.19: 10570478 - 10577386 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00511710_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem138
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK013694.1 - 2 - 3 464 65
          AK031921.1 - 3 - 4 401 65
          AK050471.1 - 3 - 4 467 65
          BC058237.1 - 3 - 4 728 6307765 65
          BC107026.2 - 2 - 3 436 40050148 65
          BC132574.1 - 2 - 3 330 40130851 65
          BC132578.1 - 2 - 3 330 40130855 65
          BC145197.1 - 2 - 3 257 65

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31951461 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31951460 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31951462 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P399140 BC145197.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P399140 AK031921.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P399140 AK050471.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P399140 BC058237.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P399140 BC107026.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P399140 BC145197.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P399140 AK031921.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P399140 AK050471.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P399140 BC058237.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P399140 BC107026.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 403338 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 724623 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 92650 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 282208 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 60917 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P139023 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P399140 BC145197.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P399140 AK031921.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P399140 AK050471.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P399140 BC058237.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P399140 BC107026.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P399140 BC145197.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P399140 AK031921.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P399140 AK050471.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P399140 BC058237.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P399140 BC107026.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31951456 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31951430 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31951457 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31951451 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31951433 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31951439 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31951429 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31951437 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31951459 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31951460 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31951454 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31951444 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31951447 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31951440 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31951432 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31951462 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31951436 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31951438 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31951461 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31951442 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31951463 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31951464 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31951449 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31951445 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P139023 - -
          Whole Mouse Genome G4122A A_52_P399140 BC145197.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P399140 AK031921.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P399140 AK050471.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P399140 BC058237.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P399140 BC107026.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P399140 BC145197.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P399140 AK031921.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P399140 AK050471.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P399140 BC058237.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P399140 BC107026.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120296 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P139023 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14355992 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14355989 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14355981 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31951462 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31951461 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31951460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31951410 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14507893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14382857 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31951427 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31951422 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31951417 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31951434 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31951421 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14392738 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31951430 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31951459 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31951457 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31951415 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31951447 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31951438 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31951454 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31951440 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31951461 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14272073 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31951456 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31951424 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31951460 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14507898 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31951449 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31951450 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31951433 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31951444 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31951437 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14456066 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31951420 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31951445 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10466003 - -
          Affymetrix Mouse Gene 1.0 ST Array 10466001 - -
          Affymetrix Mouse Gene 1.0 ST Array 10466006 - -
          Affymetrix Mouse Gene 1.0 ST Array 10466002 - -
          Affymetrix Mouse Gene 1.0 ST Array 10466007 - -
          Affymetrix Mouse Gene 1.0 ST Array 10466004 - -
          Affymetrix Mouse Gene 1.0 ST Array 10466005 - -
          Affymetrix HT MG-430 PM Array Plate 1453831_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1452978_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10466000 - -
          Mouse Genome 430 2.0 Array 1453831_PM_at - -
          Mouse Genome 430 2.0 Array 1452978_at - -
          Mouse Genome 430 2.0 Array 1452978_PM_at - -
          Mouse Genome 430 2.0 Array 1453831_at - -
          Mouse Expression Set 430 1453831_PM_at - -
          Mouse Expression Set 430 1452978_PM_at - -
          Mouse Expression Set 430 1453831_at - -
          Mouse Expression Set 430 1452978_at - -
          Affymetrix HT Mouse Genome 430B Array 1452978_at - -
          Affymetrix HT Mouse Genome 430B Array 1452978_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453831_at - -
          Affymetrix HT Mouse Genome 430B Array 1453831_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 540048 - -
          Illumina mouse-6 v1.0 expression beadchip 540048 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000540048 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2610554 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2610554 - -
          mouse-6 v1.1 (Illumina) scl52748.5.1_9-S - -
          Illumina Sentrix mouse-6.v1 bead array 540048 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl52748.5.1_9-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 540048 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2610554 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2610554 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline