Search
Search

Calr3| Gene Symbol: Calr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028500.3 | NP_082776.2 | 2 - 3 | 284 | 87 | |
| GenBank mRNA | AB206786.1 | - | 2 - 3 | 189 | 87 | |
| AK006582.1 | - | 2 - 3 | 285 | 87 | ||
Set Membership: |
|