Search
Search

4930447C04Rik| Gene Symbol: 4930447C04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029444.2 | NP_083720.1 | 6 - 7 | 579 | 90 | |
| GenBank mRNA | AK014811.1 | - | 6 - 7 | 592 | 90 | |
| AK015397.1 | - | 6 - 7 | 579 | 90 | ||
| BC137875.1 | - | 7 - 8 | 654 | 8860894 | 90 | |
Set Membership: |
|