Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00513052_m1
          See other FLYWCH2 GE Assays ›
          Gene Symbol:
          Flywch2
          Gene Name:
          FLYWCH family member 2
          Gene Aliases:
          2810417J12Rik, 3010014L17Rik, AW548203
          Chromosome Location:
          Chr.17: 23776918 - 23786077 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00513052_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Flywch2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029798.3 NP_084074.1 2 - 3 502 63
          GenBank mRNA AK013108.1 - 2 - 3 504 63
          AK019378.1 - 2 - 3 490 63
          BC024947.1 - 2 - 3 458 2648415 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700001605.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Flywch2

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31093653 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31093651 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31093649 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31093652 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31093650 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31093668 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31093670 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P500541 BC024947.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P500541 NM_029798.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P500541 AK013108.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P500541 AK019378.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P500541 BC024947.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P500541 NM_029798.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P500541 AK013108.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P500541 AK019378.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 870268 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 51185 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 290249 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 52361 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 902691 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 792613 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 902165 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 98917 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P166679 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P500541 BC024947.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P500541 NM_029798.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P500541 AK013108.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P500541 AK019378.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P500541 BC024947.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P500541 NM_029798.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P500541 AK013108.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P500541 AK019378.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093652 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093651 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093649 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093638 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093663 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093640 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093674 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093643 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093647 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093645 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093654 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093655 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093667 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093670 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093648 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093642 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P166679 - -
          Whole Mouse Genome G4122A A_51_P500541 BC024947.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P500541 NM_029798.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P500541 AK013108.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P500541 AK019378.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P500541 BC024947.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P500541 NM_029798.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P500541 AK013108.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P500541 AK019378.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113913 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109812 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P166679 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13522770 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13066057 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13522772 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13522771 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13525487 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13066051 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31093649 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31093650 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31093653 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31093652 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31093670 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31093668 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31093651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093666 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093645 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13522772 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093619 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093628 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13066056 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093581 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13232854 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093658 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093678 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13312045 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093583 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13273071 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093593 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093598 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13490601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13295822 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093660 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13222469 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093672 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13470359 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093687 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13480604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093681 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13385649 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093617 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13222470 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13295804 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093591 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13295796 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093659 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093679 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13522775 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093637 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13398883 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093632 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093661 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093615 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093663 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093675 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093648 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13445417 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093653 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13295824 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093671 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093684 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13557574 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093674 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093599 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093670 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093588 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093641 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093613 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10448377 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448379 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448378 - -
          Affymetrix HT MG-430 PM Array Plate 1424494_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1438982_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10448376 - -
          Mouse Genome 430 2.0 Array 1438982_PM_s_at - -
          Mouse Genome 430 2.0 Array 1424494_PM_s_at - -
          Mouse Genome 430 2.0 Array 1438982_s_at - -
          Mouse Genome 430 2.0 Array 1424494_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76917_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76917_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700014062.mm.1 NM_029798.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014062.mm.1 NM_029798.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014062.mm.1 TC1700001605.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700014059.mm.1 NM_029798.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014059.mm.1 NM_029798.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014059.mm.1 TC1700001605.mm.1 MTA TC
          Mouse Expression Set 430 1424494_s_at - -
          Mouse Expression Set 430 1438982_PM_s_at - -
          Mouse Expression Set 430 1438982_s_at - -
          Mouse Expression Set 430 1424494_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1424494_s_at - -
          Mouse Genome 430A 2.0 Array 1438982_s_at - -
          Mouse Genome 430A 2.0 Array 1438982_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1424494_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1424494_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1438982_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1438982_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1424494_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 333472 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3360152 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003360152 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1256528 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1256528 - -
          mouse-6 v1.1 (Illumina) scl50230.5.1_81-S - -
          Illumina Sentrix mouse-6.v1 bead array 3360152 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl50230.5.1_81-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3360152 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1256528 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1256528 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl50230.5.1_81-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline