Search
Search

Arfip2| Gene Symbol: Arfip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029802.4 | NP_084078.3 | 6 - 7 | 908 | 80 | |
| GenBank mRNA | AK009100.1 | - | 6 - 7 | 846 | 80 | |
| AK154772.1 | - | 6 - 7 | 874 | 80 | ||
| BC013794.1 | - | 6 - 7 | 819 | 2648132 | 80 | |
| BC022942.1 | - | 6 - 7 | 832 | 3583149 | 80 | |
| BC028310.1 | - | 6 - 7 | 823 | 80 | ||
| BC034520.1 | - | 6 - 7 | 889 | 3964827 | 80 | |
| BC079603.1 | - | 3 - 4 | 2081 | 80 | ||
| BU583325.1 | - | 2 - 3 | 363 | 80 | ||
Set Membership: |
|