Search
Search
| Gene Symbol: Dlst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030225.4 | NP_084501.1 | 6 - 7 | 400 | 87 | |
| GenBank mRNA | AK019713.2 | - | 397 | 87 | ||
| AK054053.1 | - | 6 - 7 | 354 | 87 | ||
| AK149664.1 | - | 6 - 7 | 376 | 87 | ||
| AK158877.1 | - | 6 - 7 | 401 | 87 | ||
| AK168570.1 | - | 6 - 7 | 376 | 87 | ||
| AK169943.1 | - | 6 - 7 | 354 | 87 | ||
| BC006702.1 | - | 6 - 7 | 337 | 3583936 | 87 | |
| CT010197.1 | - | 6 - 7 | 336 | 87 | ||
Set Membership: |
|