Search
Search
| Gene Symbol: Comt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111062.1 | NP_001104532.1 | 3 - 4 | 690 | 66 | |
| NM_001111063.1 | NP_001104533.1 | 2 - 3 | 501 | 66 | ||
| NM_007744.3 | NP_031770.2 | 2 - 3 | 633 | 66 | ||
| GenBank mRNA | AF076156.1 | - | 2 - 3 | 435 | 66 | |
| AK148311.1 | - | 3 - 4 | 357 | 66 | ||
| BC010402.1 | - | 2 - 3 | 427 | 4210097 | 66 | |
| CB946852.1 | - | 2 - 3 | 470 | 66 | ||
| CX237493.1 | - | 2 - 3 | 633 | 66 | ||
Set Membership: |
|