Search
Search
| Gene Symbol: Emd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007927.3 | NP_031953.1 | 1 - 2 | 350 | 74 | |
| GenBank mRNA | AK146361.1 | - | 1 - 2 | 196 | 74 | |
| AK160531.1 | - | 1 - 2 | 195 | 74 | ||
| AK167852.1 | - | 1 - 2 | 193 | 74 | ||
| BC132131.1 | - | 1 - 2 | 162 | 40130408 | 74 | |
| BC132133.1 | - | 1 - 2 | 162 | 40130410 | 74 | |
| BY260524.1 | - | 1 - 2 | 351 | 74 | ||
| U73902.1 | - | 1 - 2 | 200 | 74 | ||
Set Membership: |
|