Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mecom | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007963.2 | NP_031989.2 | 14 - 15 | 2876 | 81 | |
GenBank mRNA | AK033517.1 | - | 14 - 15 | 2310 | 81 | |
BC076620.1 | - | 14 - 15 | 2828 | 81 | ||
JQ665270.1 | - | 12 - 13 | 2996 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |