Search
Search
| Gene Symbol: Fbn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007993.2 | NP_032019.2 | 21 - 22 | 2831 | 71 | |
| GenBank mRNA | AF007248.1 | - | 2545 | 71 | ||
| L29454.1 | - | 4918 | 71 | |||
| U19972.1 | - | 20 - 21 | 2471 | 71 | ||
| U22493.1 | - | 21 - 22 | 2764 | 71 | ||
Set Membership: |
|